(S)-(-)-2-Chloropropionic acid
Supplier: Apollo Scientific
Danger
Synonyms: (S)-(-)-2-Chloropropanoic acid
OR28944-100G OR28944-25G OR28944-5G
APOSOR28944-100GEA 119 EUR
APOSOR28944-100G APOSOR28944-25G APOSOR28944-5G
(S)-(-)-2-Chloropropionic acid
(S)-(-)-2-Chloropropionic acid
Formula: H₃CCHClCOOH
MW: 108.52 g/mol
Boiling Pt: 77 °C/10mmHg
Melting Pt: 4 °C
Density: 1.249 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00064205
CAS Number: 29617-66-1
EINECS: 411-150-2
UN: 2511
ADR: 8 III