2-Methylimidazole 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Methyl-1H-imidazole
OR2900-100G OR2900-25G OR2900-500G
APOSOR2900-100GEA 37.7 EUR
APOSOR2900-100G APOSOR2900-25G APOSOR2900-500G
2-Methylimidazole 98%
2-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 267…268 °C
Melting Pt: 142…144 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 1759
ADR: 8,III