3-(1,3-Oxazol-5-yl)aniline
Supplier: Apollo Scientific
Warning
Synonyms: 3-(5-Oxazolyl)aniline
OR29046-1G OR29046-250MG
APOSOR29046-1GEA 34.8 EUR
APOSOR29046-1G APOSOR29046-250MG
3-(1,3-Oxazol-5-yl)aniline
3-(1,3-Oxazol-5-yl)aniline
Formula: C₉H₈N₂O
MW: 160.1700 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00052191
CAS Number: 157837-31-5