3-Phenylbut-2-enoic acid
Supplier: Apollo Scientific
Warning
OR29067-1G OR29067-250MG OR29067-500MG
APOSOR29067-1GEA 451 EUR
APOSOR29067-1G APOSOR29067-250MG APOSOR29067-500MG
3-Phenylbut-2-enoic acid
3-Phenylbut-2-enoic acid
Formula: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00053500
CAS Number: 1199-20-8