1-(2,4-Dimethyl-1,3-oxazol-5-yl)ethanone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Acetyl-2,4-dimethyl-1,3-oxazole
OR29075-1G
OR29075-250MG
APOSOR29075-1GEA
134
EUR
APOSOR29075-1G
APOSOR29075-250MG
1-(2,4-Dimethyl-1,3-oxazol-5-yl)ethanone
1-(2,4-Dimethyl-1,3-oxazol-5-yl)ethanone
Formula:
C₇H₉NO₂ MW: 139.16 g/mol Melting Pt: 56…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD01934451 CAS Number: 23012-25-1 |