3-Bromothiophene-2-carbaldehyde
Supplier: Apollo Scientific
Warning
OR29108-1G OR29108-25G OR29108-5G
APOSOR29108-1GEA 21.9 EUR
APOSOR29108-1G APOSOR29108-25G APOSOR29108-5G
3-Bromothiophene-2-carbaldehyde
3-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
Boiling Pt: 110…113 °C/10mmHg
Melting Pt: 24…25 °C
Density: 1.755 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00126680
CAS Number: 930-96-1