3-(Chloromethyl)-5-(2-methoxyphenyl)-1,2,4-oxadiazole
Supplier: Apollo Scientific
Danger
OR29122-1G OR29122-5G
APOSOR29122-1GEA 92.8 EUR
APOSOR29122-1G APOSOR29122-5G
3-(Chloromethyl)-5-(2-methoxyphenyl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(2-methoxyphenyl)-1,2,4-oxadiazole
Formula: C₁₀H₉ClN₂O₂
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00084965
CAS Number: 175205-61-5
UN: 3261
ADR: 8 III