3-Cyanobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Formylbenzonitrile
OR29140-25G OR29140-5G OR29140-100G
APOSOR29140-25GEA 27.7 EUR
APOSOR29140-25G APOSOR29140-5G APOSOR29140-100G
3-Cyanobenzaldehyde 98%
3-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 209…210 °C
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00003344
CAS Number: 24964-64-5
EINECS: 246-549-1