3,5-Dibromobenzonitrile
Supplier: Apollo Scientific
Warning
OR29247-25G OR29247-5G
APOSOR29247-25GEA 79.9 EUR
APOSOR29247-25G APOSOR29247-5G
3,5-Dibromobenzonitrile
3,5-Dibromobenzonitrile
Formula: C₇H₃Br₂N
MW: 260.9134 g/mol
Melting Pt: 98.5…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00178770
CAS Number: 97165-77-0
UN: 3439
ADR: 6.1,III