2-(4-Hydroxyphenyl)ethanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
OR2934-25G OR2934-5G OR2934-100G
APOSOR2934-25GEA 26.1 EUR
APOSOR2934-25G APOSOR2934-5G APOSOR2934-100G
2-(4-Hydroxyphenyl)ethanol 98%
2-(4-Hydroxyphenyl)ethanol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 139 °C/1mmHg
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8