Ethyl pyrrole-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 1H-pyrrole-2-carboxylate
OR29376-5G OR29376-1G
APOSOR29376-5GEA 24.8 EUR
APOSOR29376-5G APOSOR29376-1G
Ethyl pyrrole-2-carboxylate
Ethyl pyrrole-2-carboxylate
Formula: C₇H₉NO₂
MW: 139.16 g/mol
Boiling Pt: 70 °C/0.8mmHg
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00817049
CAS Number: 2199-43-1