ω-Bromo-4-cyanoacetophenone 95%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone
OR29653-100G OR29653-25G OR29653-5G
APOSOR29653-100GEA 328 EUR
APOSOR29653-100G APOSOR29653-25G APOSOR29653-5G
ω-Bromo-4-cyanoacetophenone 95%
ω-Bromo-4-cyanoacetophenone
Formula: C₉H₆BrNO
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00052931
CAS Number: 20099-89-2
UN: 2923
ADR: 8+6.1,II