(2,6-Dichlorophenoxy)acetonitrile
Supplier: Apollo Scientific
OR29660-1G OR29660-250MG OR29660-500MG
APOSOR29660-1GEA 256 EUR
APOSOR29660-1G APOSOR29660-250MG APOSOR29660-500MG
(2,6-Dichlorophenoxy)acetonitrile
(2,6-Dichlorophenoxy)acetonitrile
Formula: C₈H₅Cl₂NO
Storage Temperature: Ambient
CAS Number: 21244-78-0