4-Chloro-2-methylbenzylamine
Supplier: Apollo Scientific
Danger
OR2971-1G OR2971-5G
APOSOR2971-1GEA 66.7 EUR
APOSOR2971-1G APOSOR2971-5G
4-Chloro-2-methylbenzylamine
4-Chloro-2-methylbenzylamine
Formula: C₈H₁₀ClN
Storage Temperature: Ambient
MDL Number: MFCD00047932
CAS Number: 27917-11-9
UN: 3259
ADR: 8 III