(S)-(-)-1-(p-Tolyl)ethylamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: (S)-(-)-4-(1-Aminoethyl)toluene, (S)-1-(p-Tolyl)ethanamine
OR2973-25G OR2973-5G
APOSOR2973-25GEA 0 EUR
APOSOR2973-25G APOSOR2973-5G
(S)-(-)-1-(p-Tolyl)ethylamine 98%
(S)-(-)-1-(p-Tolyl)ethylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 204 °C
Melting Pt: < –20 °C
Density: 0.94 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00145246
CAS Number: 27298-98-2
UN: 2735
ADR: 8 III