6-Chloronicotinonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-5-cyanopyridine, 6-Chloro-3-pyridinecarbonitrile, 2-Chloropyridine-5-carbonitrile
OR29741-100G OR29741-25G OR29741-5G
APOSOR29741-100GEA 114 EUR
APOSOR29741-100G APOSOR29741-25G APOSOR29741-5G
6-Chloronicotinonitrile
6-Chloronicotinonitrile
Formula: C₆H₃ClN₂
Boiling Pt: 105…107 °C/1mmHg
Melting Pt: 113…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00084941
CAS Number: 33252-28-7
UN: 3439
ADR: 6.1,III