5-Cyano-2-methylpyridine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 6-Methylnicotinonitrile, 5-Cyano-2-picoline
OR29756-25G OR29756-5G OR29756-1G
APOSOR29756-25GEA 78.3 EUR
APOSOR29756-25G APOSOR29756-5G APOSOR29756-1G
5-Cyano-2-methylpyridine 98%
5-Cyano-2-methylpyridine
Formula: C₇H₆N₂
Boiling Pt: 216…217 °C
Melting Pt: 83…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038042
CAS Number: 3222-48-8
UN: 2811
ADR: 6.1,III