Ethyl-6-chloronicotinate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 6-chloronicotinate
OR29759-100G OR29759-25G
APOSOR29759-100GEA 60.9 EUR
APOSOR29759-100G APOSOR29759-25G
Ethyl-6-chloronicotinate 97%
Ethyl 6-chloronicotinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 86…87 °C/1mmHg
Melting Pt: 26…30 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00082739
CAS Number: 49608-01-7