5-Bromo-4-methyl-2-phenyl-1,3-thiazole
Supplier: Apollo Scientific
Warning
OR2985-10G OR2985-1G OR2985-5G
APOSOR2985-10GEA 406 EUR
APOSOR2985-10G APOSOR2985-1G APOSOR2985-5G
5-Bromo-4-methyl-2-phenyl-1,3-thiazole
5-Bromo-4-methyl-2-phenyl-1,3-thiazole
Formula: C₁₀H₈BrNS
Melting Pt: 60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974049
CAS Number: 28771-82-6