Thiophene-2-carboxamidine hydrochloride
Supplier: Apollo Scientific
Warning
OR2996-1G OR2996-5G
APOSOR2996-1GEA 72.5 EUR
APOSOR2996-1G APOSOR2996-5G
Thiophene-2-carboxamidine hydrochloride
Thiophene-2-carboxamidine hydrochloride
Formula: C₅H₇ClN₂S
Melting Pt: 173…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00173786
CAS Number: 54610-70-7