5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one
Supplier: Apollo Scientific
Warning
Synonyms: 1-Oxoisoindoline-5-boronic acid, pinacol ester
OR300008-1G OR300008-250MG OR300008-5G
APOSOR300008-1GEA 219 EUR
APOSOR300008-1G APOSOR300008-250MG APOSOR300008-5G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoindolin-1-one
Formula: C₁₄H₁₈BNO₃
MW: 259.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11040393
CAS Number: 376584-62-2