5-Chloropyrido[3,4-b]pyrazine
Supplier: Apollo Scientific
OR300014-1G OR300014-250MG OR300014-500MG
APOSOR300014-1GEA 1140 EUR
APOSOR300014-1G APOSOR300014-250MG APOSOR300014-500MG
5-Chloropyrido[3,4-b]pyrazine
5-Chloropyrido[3,4-b]pyrazine
Formula: C7H4ClN3
MW: 165.5800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706209
CAS Number: 214045-82-6