2-Cyano-5-hydroxymethylpyridine
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Hydroxymethyl)picolinonitrile, 5-(Hydroxymethyl)pyridine-2-carbonitrile
OR300195-1G OR300195-250MG
APOSOR300195-1GEA 156 EUR
APOSOR300195-1G APOSOR300195-250MG
2-Cyano-5-hydroxymethylpyridine
2-Cyano-5-hydroxymethylpyridine
Formula: C₇H₆N₂O
MW: 134.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09029606
CAS Number: 58553-48-3