Ethyl-4,8-dichloroquinoline-3-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4,8-dichloroquinoline-3-carboxylate
OR300367-1G OR300367-250MG
APOSOR300367-1GEA 78.3 EUR
APOSOR300367-1G APOSOR300367-250MG
Ethyl-4,8-dichloroquinoline-3-carboxylate
Ethyl-4,8-dichloroquinoline-3-carboxylate
Formula: C₁₂H₉Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD00173351
CAS Number: 56824-91-0