3-Bromo-5-chloro-1H-pyrrolo[3,2-b]pyridine
Supplier: Apollo Scientific
Warning
OR300385-1G OR300385-250MG
APOSOR300385-1GEA 658 EUR
APOSOR300385-1G APOSOR300385-250MG
3-Bromo-5-chloro-1H-pyrrolo[3,2-b]pyridine
3-Bromo-5-chloro-1H-pyrrolo[3,2-b]pyridine
Formula: C7H4BrClN2
MW: 231.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12962966
CAS Number: 1190310-75-8