2-Hydroxy-4-methylnicotinic acid
Supplier: Apollo Scientific
Warning
OR300502-1G OR300502-500MG
APOSOR300502-1GEA 255 EUR
APOSOR300502-1G APOSOR300502-500MG
2-Hydroxy-4-methylnicotinic acid
2-Hydroxy-4-methylnicotinic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 247…250 °C
Storage Temperature: Ambient
MDL Number: MFCD09033815
CAS Number: 38076-81-2