3-Bromo-5-methylpyridin-2-ylamine hydrobromide
Supplier: Apollo Scientific
Warning
OR300591-1G OR300591-5G
APOSOR300591-1GEA 0 EUR
APOSOR300591-1G APOSOR300591-5G
3-Bromo-5-methylpyridin-2-ylamine hydrobromide
3-Bromo-5-methylpyridin-2-ylamine hydrobromide
Formula: C₆H₈Br₂N₂
MW: 267.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384840
CAS Number: 1299607-37-6