Ethyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 4-oxotetrahydro-2H-thiopyran-3-carboxylate
OR300631-1G OR300631-250MG
APOSOR300631-1GEA 169 EUR
APOSOR300631-1G APOSOR300631-250MG
Ethyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate
Ethyl-4-oxotetrahydro-2H-thiopyran-3-carboxylate
Formula: C₈H₁₂O₃S
MW: 188.25 g/mol
Boiling Pt: 97…99 °C/0.1mmHg
Melting Pt: 58…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00040797
CAS Number: 1198-44-3