Ethyl-1-bromocyclobutanecarboxylate 97%
Supplier: Apollo Scientific
Danger
OR300697-1G OR300697-25G OR300697-5G
APOSOR300697-1GEA 21.9 EUR
APOSOR300697-1G APOSOR300697-25G APOSOR300697-5G
Ethyl-1-bromocyclobutanecarboxylate 97%
Ethyl-1-bromocyclobutanecarboxylate
Formula: C₇H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00001322
CAS Number: 35120-18-4
UN: 3265
ADR: 8,III