1-Ethyl-2-iodobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Ethyliodobenzene, 1-Iodo-2-ethylbenzene
OR30075-100G OR30075-25G
APOSOR30075-100GEA 49.3 EUR
APOSOR30075-100G APOSOR30075-25G
1-Ethyl-2-iodobenzene
1-Ethyl-2-iodobenzene
Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 221 °C
Density: 1.618 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00040871
CAS Number: 18282-40-1