1-(4-Chloropyridin-2-yl)ethanone 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Chloropyridine-2-yl)ethanone, 2-Acetyl-4-chloropyridine
OR300751-1G OR300751-250MG
APOSOR300751-1GEA 82.8 EUR
APOSOR300751-1G APOSOR300751-250MG
1-(4-Chloropyridin-2-yl)ethanone 99%
1-(4-Chloropyridin-2-yl)ethanone
Formula: C₇H₆ClNO
MW: 155.58 g/mol
Melting Pt: 37…38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697662
CAS Number: 60159-37-7