2-Cyano-3-methyl-but-2-enoic acid ethyl ester
Supplier: Apollo Scientific
Synonyms: Ethyl 2-cyano-3-methylbut-2-enoate
OR300769-1G OR300769-250MG OR300769-5G OR300769-10G OR300769-25G
APOSOR300769-1GEA 52.2 EUR
APOSOR300769-1G APOSOR300769-250MG APOSOR300769-5G APOSOR300769-10G APOSOR300769-25G
2-Cyano-3-methyl-but-2-enoic acid ethyl ester
2-Cyano-3-methyl-but-2-enoic acid ethyl ester
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009135
CAS Number: 759-58-0