4-Bromo-2-benzofuran-1[3H]-one 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromophthalide
OR301006-1G OR301006-250MG
APOSOR301006-1GEA 133 EUR
APOSOR301006-1G APOSOR301006-250MG
4-Bromo-2-benzofuran-1[3H]-one 97%
4-Bromo-2-benzofuran-1[3H]-one
Formula: C₈H₅BrO₂
MW: 213.03 g/mol
Boiling Pt: 378 °C
Melting Pt: 98…104 °C
Storage Temperature: Ambient
MDL Number: MFCD06412577
CAS Number: 102308-43-0