Methyl-6-chloropyrazine-2-carboxylate 97+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 6-chloropyrazine-2-carboxylate
OR301016-5G OR301016-25G
APOSOR301016-5GEA 92.8 EUR
APOSOR301016-5G APOSOR301016-25G
Methyl-6-chloropyrazine-2-carboxylate 97+%
Methyl-6-chloropyrazine-2-carboxylate
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 40…45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10686598
CAS Number: 23611-75-8