4-Acetylbenzonitrile
Supplier: Apollo Scientific
Warning
Synonyms: Methyl p-cyanophenyl ketone, 4'-Cyanoacetophenone, 4-Acetylbenzenecarbonitrile
OR30102-100G OR30102-25G OR30102-5G
APOSOR30102-100GEA 173 EUR
APOSOR30102-100G APOSOR30102-25G APOSOR30102-5G
4-Acetylbenzonitrile
4-Acetylbenzonitrile
Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 95…96 °C/14mmHg
Melting Pt: 55…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001825
CAS Number: 1443-80-7
EINECS: 215-885-0
UN: 3439
ADR: 6.1,III