1,3-Dimethylimidazolium iodide
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Dimethyl-1H-imidazol-3-ium iodide
OR301041-5G OR301041-25G
APOSOR301041-5GEA 21.9 EUR
APOSOR301041-5G APOSOR301041-25G
1,3-Dimethylimidazolium iodide
1,3-Dimethylimidazolium iodide
Formula: C₅H₉IN₂
MW: 224.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159685
CAS Number: 4333-62-4