3'-Hydroxy-4'-methoxyacetophenone
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxy-4-methoxyacetophenone
OR301055-1G OR301055-25G OR301055-5G
APOSOR301055-1GEA 21.9 EUR
APOSOR301055-1G APOSOR301055-25G APOSOR301055-5G
3'-Hydroxy-4'-methoxyacetophenone
3'-Hydroxy-4'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00182975
CAS Number: 6100-74-9