2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine 98%
Supplier: Apollo Scientific
Warning
OR301065-25G
APOSOR301065-25GEA 150 EUR
APOSOR301065-25G
2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine 98%
2,4-Dichloro-5H-pyrrolo[3,2-d]pyrimidine
Formula: C₆H₃Cl₂N₃
MW: 188.0141 g/mol
Melting Pt: 229…231 °C
Storage Temperature: Ambient
MDL Number: MFCD08059281
CAS Number: 63200-54-4