2,3-Dimethylbenzoyl chloride
Supplier: Apollo Scientific
Danger
OR30107-25G OR30107-5G OR30107-1G
APOSOR30107-25GEA 328 EUR
APOSOR30107-25G APOSOR30107-5G APOSOR30107-1G
2,3-Dimethylbenzoyl chloride
2,3-Dimethylbenzoyl chloride
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 122…124 °C
Melting Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00045217
CAS Number: 21900-46-9
UN: 3265
ADR: 8,II