3-Bromo-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR301082-10G OR301082-1G OR301082-5G
APOSOR301082-10GEA 87 EUR
APOSOR301082-10G APOSOR301082-1G APOSOR301082-5G
3-Bromo-1H-pyrazole
3-Bromo-1H-pyrazole
Formula: C3H3BrN2
MW: 146.97 g/mol
Boiling Pt: 70…72 °C/0.1mmHg
Melting Pt: 60…65 °C
Storage Temperature: Freezer
MDL Number: MFCD01646049
CAS Number: 14521-80-3