4-Bromo-3-hydroxypyridine 95%
Supplier: Apollo Scientific
Warning
OR301092-10G OR301092-1G OR301092-5G
APOSOR301092-10GEA 325 EUR
APOSOR301092-10G APOSOR301092-1G APOSOR301092-5G
4-Bromo-3-hydroxypyridine 95%
4-Bromo-3-hydroxypyridine
Formula: C₅H₄BrNO
MW: 173.9954 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD03093021
CAS Number: 161417-28-3