Methyl 5-hydroxypyrazine-2-carboxylate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-hydroxypyrazine-2-carboxylate
OR301122-10G OR301122-25G OR301122-5G
APOSOR301122-10GEA 137 EUR
APOSOR301122-10G APOSOR301122-25G APOSOR301122-5G
Methyl 5-hydroxypyrazine-2-carboxylate 95%
Methyl-5-hydroxypyrazine-2-carboxylate
Formula: C₆H₆N₂O₃
MW: 154.1234 g/mol
Melting Pt: 181…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08273469
CAS Number: 13924-95-3