4-(4-Bromophenyl)butyric acid 97%
Supplier: Apollo Scientific
Synonyms:
4-(4-Bromophenyl)butanoic acid
OR301143-5G
APOSOR301143-5GEA
53.9
EUR
APOSOR301143-5G
4-(4-Bromophenyl)butyric acid 97%
4-(4-Bromophenyl)butyric acid
Formula:
C₁₀H₁₁BrO₂ MW: 243.10 g/mol Melting Pt: 71…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00463170 CAS Number: 35656-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...