7-Chloro-1H-pyrazolo[3,4-c]pyridine
Supplier: Apollo Scientific
Warning
OR301174-1G OR301174-5G
APOSOR301174-1GEA 45.1 EUR
APOSOR301174-1G APOSOR301174-5G
7-Chloro-1H-pyrazolo[3,4-c]pyridine
7-Chloro-1H-pyrazolo[3,4-c]pyridine
Formula: C₆H₄ClN₃
MW: 153.5691 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265710
CAS Number: 76006-11-6