Morpholin-3-one
Supplier: Apollo Scientific
Danger
Synonyms: 3-Morpholinone
OR301177-100G OR301177-25G OR301177-5G
APOSOR301177-100GEA 109 EUR
APOSOR301177-100G APOSOR301177-25G APOSOR301177-5G
Morpholin-3-one
Morpholin-3-one
Formula: C₄H₇NO₂
MW: 101.1039 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00631009
CAS Number: 109-11-5
EINECS: 203-647-9