4-Chloro-1H-pyrazolo[3,4-b]pyridine
Supplier: Apollo Scientific
Warning
OR301233-1G OR301233-5G OR301233-25G
APOSOR301233-1GEA 85.7 EUR
APOSOR301233-1G APOSOR301233-5G APOSOR301233-25G
4-Chloro-1H-pyrazolo[3,4-b]pyridine
4-Chloro-1H-pyrazolo[3,4-b]pyridine
Formula: C₆H₄ClN₃
MW: 153.569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846313
CAS Number: 29274-28-0