6-Chloro-2-methylpyrimidin-4-amine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-6-chloro-2-methylpyrimidine, 4-Pyrimidinamine, 6-chloro-2-methyl-, 6-chloro-2-methylpyrimidin-4-amine
OR301253-100G OR301253-25G OR301253-5G
APOSOR301253-100GEA 159 EUR
APOSOR301253-100G APOSOR301253-25G APOSOR301253-5G
6-Chloro-2-methylpyrimidin-4-amine 97%
6-Chloro-2-methylpyrimidin-4-amine
Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07782029
CAS Number: 1749-68-4