6-Hydroxypyridazine-3-carboxylic acid 98%
Supplier: Apollo Scientific
Warning
OR301257-25G
APOSOR301257-25GEA 60.9 EUR
APOSOR301257-25G
6-Hydroxypyridazine-3-carboxylic acid 98%
6-Hydroxypyridazine-3-carboxylic acid
Formula: C₅H₄N₂O₃
MW: 140.1000 g/mol
Melting Pt: 246…247 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052805
CAS Number: 37972-69-3