2,6-Dichloro-4-iodopyridine 98%
Supplier: Apollo Scientific
Danger
OR301306-100G OR301306-25G OR301306-5G
APOSOR301306-100GEA 248 EUR
APOSOR301306-100G APOSOR301306-25G APOSOR301306-5G
2,6-Dichloro-4-iodopyridine 98%
2,6-Dichloro-4-iodopyridine
Formula: C₅H₂CI₂IN
MW: 273.89 g/mol
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD07368400
CAS Number: 98027-84-0