2,4,6-Trimethylbenzonitrile
Supplier: Apollo Scientific
Warning
OR30133-25G
APOSOR30133-25GEA 191 EUR
APOSOR30133-25G
2,4,6-Trimethylbenzonitrile
2,4,6-Trimethylbenzonitrile
Formula: C₁₀H₁₁N
MW: 145.2 g/mol
Boiling Pt: 79…80 °C/2mmHg
Melting Pt: 50…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00016378
CAS Number: 2571-52-0